Novus Biologicals
Manufacturer Code:NBP258532
Catalog # NBP258532
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LTPRKRKQDSLKCDSLLHFTENLFPSPNKKHCFYQNSDKNEENLHCSQQEHFVLSALKTTEINRLPSANQGSPFKSALSTVSF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2410004I17Rik EC 2.3.1.- ECO1 homolog 2 EFO2 Establishment of cohesion 1 homolog 2 establishment of cohesion 1 homolog 2 (S. cerevisiae) N-acetyltransferase ESCO2 RBS Roberts syndrome; ECO1 homolog 2; EFO2; Establishment factor-like protein 2; Establishment of cohesion 1 homolog 2; N-acetyltransferase ESCO2
Gene Aliases: 2410004I17Rik; EFO2; ESCO2; RBS
UniProt ID: (Human) Q56NI9
Entrez Gene ID: (Human) 157570
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.