Proteintech
Manufacturer Code:235251AP150UL
Catalog # 50-173-4089
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ESCO2 Fusion Protein Ag18893 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2410004I17Rik ECO1 homolog 2 EFO2 ESCO2 N acetyltransferase ESCO2 RBS; ECO1 homolog 2; EFO2; Establishment factor-like protein 2; Establishment of cohesion 1 homolog 2; N-acetyltransferase ESCO2
Gene Aliases: 2410004I17Rik; D030072L07Rik; EFO2; ESCO2; RBS
UniProt ID: (Human) Q56NI9, (Mouse) Q8CIB9
Entrez Gene ID: (Human) 157570, (Mouse) 71988
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.