Invitrogen
Manufacturer Code:PA537183
Catalog # PIPA537183
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues in human ESCO1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CTF7 homolog 1; ECO1 homolog 1; EFO1; EFO1p; ESO1 homolog 1; Establishment factor-like protein 1; Establishment of cohesion 1 homolog 1; establisment of cohesion 1 homolog 1; hEFO1; N-acetyltransferase ESCO1; N-acetyltransferase ESCO1 CTF7 homolog 1 Establishment factor-like protein 1 EFO1p hEFO1 Establishment of cohesion 1 homolog 1 ECO1 homolog 1 ESO1 homolog 1 EFO1 KIAA1911; N-acetyltransferase ESCO1 variant 2
Gene Aliases: A930014I12Rik; CTF; ECO1; EFO1; ESCO1; ESO1; KIAA1911
UniProt ID: (Human) Q5FWF5, (Mouse) Q69Z69
Entrez Gene ID: (Human) 114799, (Mouse) 77805, (Rat) 680014
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.