Invitrogen
Manufacturer Code:PA5103942
Catalog # PIPA5103942
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ESCO1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A930014I12Rik CTF CTF7 homolog 1 ECO1 ECO1 homolog 1 EFO1 EFO1p Esco1 ESO1 ESO1 homolog 1 establishment factor-like protein 1 Establishment of cohesion 1 homolog 1 establishment of cohesion 1 homolog 1 (S. cerevisiae) establishment of sister chromatid cohesion N-acetyltransferase 1 establisment of cohesion 1 homolog 1 hEFO1 Kiaa1911 N-acetyltransferase ESCO1 N-acetyltransferase ESCO1 variant 2; CTF7 homolog 1; ECO1 homolog 1; EFO1; EFO1p; ESO1 homolog 1; Establishment factor-like protein 1; Establishment of cohesion 1 homolog 1; establisment of cohesion 1 homolog 1; hEFO1; N-acetyltransferase ESCO1; N-acetyltransferase ESCO1 variant 2
Gene Aliases: A930014I12Rik; CTF; ECO1; EFO1; ESCO1; ESO1; KIAA1911
UniProt ID: (Human) Q5FWF5, (Mouse) Q69Z69
Entrez Gene ID: (Human) 114799, (Mouse) 77805
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.