R&D Systems
Manufacturer Code:MAB4204SP
Catalog # MAB4204SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 408519 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human ESAM Gln30-Ala247 Accession # Q96AP7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2310008D05Rik; 2310008D05Rik endothelial cell adhesion molecule endothelial cell-selective adhesion molecule HUEL (C4orf1)-interacting protein LP4791 protein W117m; Endothelial cell-selective adhesion molecule; HUEL (C4orf1)-interacting protein; LP4791 protein
Gene Aliases: ESAM; UNQ220/PRO246; W117m
UniProt ID: (Human) Q96AP7
Entrez Gene ID: (Human) 90952
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.