Novus Biologicals
Manufacturer Code:NBP201652
Catalog # NBP201652
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2B9 |
Immunogen | Full length human recombinant protei of human PDIA4(NP_004902) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Endoplasmic reticulum resident protein 70; Endoplasmic reticulum resident protein 70 Endoplasmic reticulum resident protein 72 ER protein 70 ERp70 ERP70ER protein 72 ERp72 ERp-72 ERP72EC 5.3.4.1 intestinal-related) protein disulfide isomerase family A member 4 protein disulfide isomerase related protein (calcium-binding protein protein disulfide isomerase-associated 4 protein disulfide-isomerase A4; Endoplasmic reticulum resident protein 72; ER protein 70; ER protein 72; protein disulfide isomerase family A, member 4; protein disulfide isomerase related protein (calcium-binding protein, intestinal-related); protein disulfide isomerase-associated 4; Protein disulfide-isomerase A4; testicular tissue protein Li 137
Gene Aliases: ERp-72; ERP70; ERP72; PDIA4
UniProt ID: (Human) P13667
Entrez Gene ID: (Human) 9601
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.