Invitrogen
Manufacturer Code:PA1007A555
Catalog # PIPA1007AZ
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues K(629) F I E E H A T K L S R T K E E L(645) of humna ERp72 |
Conjugate | Alexa Fluor 555 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI987846 Erp72 ERP72 ERp-72 ERP70 Cai; calcium binding protein, intestinal; endoplasmic reticulum protein 72; Endoplasmic reticulum resident protein 70; Endoplasmic reticulum resident protein 72; ER protein 70; ER protein 72; protein disulfide isomerase family A, member 4; protein disulfide isomerase related protein (calcium-binding protein, intestinal-related); protein disulfide isomerase-associated 4; Protein disulfide-isomerase A4; testicular tissue protein Li 137
Gene Aliases: AI987846; Cai; ERp-72; ERP70; ERP72; PDIA4
UniProt ID: (Human) P13667, (Mouse) P08003
Entrez Gene ID: (Human) 9601, (Mouse) 12304
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.