Novus Biologicals
Manufacturer Code:NBP236767
Catalog # NBP236767
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL2452 |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:PTLKIFRDGEEAGAYDGPRTADGIVSHLKKQAGPASVPLRTEEEFKKFISDKDASIVGFFDDSFSEAHSEFLKAASNLRDNYRFAHTNVESLVNEYDDNGEGIILFRPSHLTNKFEDK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 58 kDa glucose-regulated protein; 58 kDa microsomal protein; Disulfide isomerase ER-60; Disulfide isomerase ER-60 EC 5.3.4.1 endoplasmic reticulum P58 Endoplasmic reticulum resident protein 57 Endoplasmic reticulum resident protein 60 ER protein 60 ER60 ERP57 ERp5758 kDa glucose-regulated protein ERP60 ERp6058 kDa microsomal protein ERp61 glucose regulated protein 58kDa GRP57 GRP58ER protein 57 HsT17083 P58 phospholipase C-alpha PI-PLC protein disulfide isomerase family A member 3 protein disulfide isomerase-associated 3 protein disulfide-isomerase A3; endoplasmic reticulum P58; Endoplasmic reticulum resident protein 57; Endoplasmic reticulum resident protein 60; epididymis secretory protein Li 269; epididymis secretory sperm binding protein Li 93n; ER protein 57; ER protein 60; glucose regulated protein, 58kDa; p58; phospholipase C-alpha; protein disulfide isomerase family A, member 3; protein disulfide isomerase-associated 3; Protein disulfide-isomerase A3
Gene Aliases: ER60; ERP57; ERP60; ERp61; GRP57; GRP58; HEL-S-269; HEL-S-93n; HsT17083; P58; PDIA3; PI-PLC
UniProt ID: (Human) P30101
Entrez Gene ID: (Human) 2923
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.