Novus Biologicals
Manufacturer Code:NBP30487020UL
Catalog # NB096158
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 223-462 of human ERRFI1 (NP_061821.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ERBB receptor feedback inhibitor 1; ERBB receptor feedback inhibitor 1 GENE-33 MIG6 MIG-6Mitogen-inducible gene 6 protein RALTreceptor-associated late transducer; MIG-6; Mitogen-inducible gene 6 protein; receptor-associated late transducer
Gene Aliases: ERRFI1; GENE-33; MIG-6; MIG6; RALT
UniProt ID: (Human) Q9UJM3
Entrez Gene ID: (Human) 54206
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.