Novus Biologicals
Manufacturer Code:NBP297000UV
Catalog # NB041607
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant ERP44 (NP_055866.1 Met1-Asp402). |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: endoplasmic reticulum protein 44 endoplasmic reticulum resident protein 44 kDa ER protein 44 ERp44 KIAA0573endoplasmic reticulum resident protein 44 PDIA10 protein disulfide isomerase family A member 10 thioredoxin domain containing 4 (endoplasmic reticulum) Thioredoxin domain-containing protein 4 TXNDC4; Endoplasmic reticulum resident protein 44; endoplasmic reticulum resident protein 44 kDa; ER protein 44; protein disulfide isomerase family A, member 10; thioredoxin domain containing 4 (endoplasmic reticulum); Thioredoxin domain-containing protein 4
Gene Aliases: ERP44; KIAA0573; PDIA10; TXNDC4; UNQ532/PRO1075
UniProt ID: (Human) Q9BS26
Entrez Gene ID: (Human) 23071
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.