Invitrogen
Manufacturer Code:MA537989
Catalog # PIMA537989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2295 |
Immunogen | A synthesized peptide derived from human ERp29 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 1200015M03Rik 2810446M09Rik AW209030 C12orf8 endoplasmic reticulum lumenal protein ERp28 endoplasmic reticulum protein 29 endoplasmic reticulum protein ERp29 endoplasmic reticulum resident protein 28 Endoplasmic reticulum resident protein 29 endoplasmic reticulum resident protein 31 endoplasmic retuclum protein 29 epididymis secretory protein Li 107 Erp28 ERp29 ERp31 HEL-S-107 PDIA9 PDI-DB protein disulfide isomerase family A member 9; endoplasmic reticulum lumenal protein ERp28; endoplasmic reticulum protein ERp29; Endoplasmic reticulum resident protein 28; Endoplasmic reticulum resident protein 29; Endoplasmic reticulum resident protein 31; endoplasmic retuclum protein 29; epididymis secretory protein Li 107; ERp28; ERp29; ERp31; protein disulfide isomerase family A, member 9
Gene Aliases: 1200015M03Rik; 2810446M09Rik; AW209030; C12orf8; ERP28; ERP29; ERp31; HEL-S-107; PDI-DB; PDIA9
UniProt ID: (Human) P30040, (Rat) P52555, (Mouse) P57759
Entrez Gene ID: (Human) 10961, (Rat) 117030, (Mouse) 67397
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.