Invitrogen
Manufacturer Code:PA519034
Catalog # PIPA519034
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (EKNKMSDGKKEELQ) corresponding to the C-terminus amino acids of ERP29 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: endoplasmic reticulum lumenal protein ERp28; endoplasmic reticulum protein ERp29; Endoplasmic reticulum resident protein 28; Endoplasmic reticulum resident protein 29; Endoplasmic reticulum resident protein 31; endoplasmic retuclum protein 29; epididymis secretory protein Li 107; ERp28; ERp29; ERP29 endoplasmic reticulum protein 29 C12orf8 ERp28 ERp31 PDI-DB endoplasmic reticulum lumenal protein ERp28; ERp31; protein disulfide isomerase family A, member 9
Gene Aliases: C12orf8; ERP28; ERP29; ERp31; HEL-S-107; PDI-DB; PDIA9
UniProt ID: (Human) P30040, (Rat) P52555
Entrez Gene ID: (Human) 10961, (Rat) 117030
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.