Novus Biologicals
Manufacturer Code:NBP299200H
Catalog # NB058906
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human ERP29. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C12orf8chromosome 12 open reading frame 8 endoplasmic reticulum lumenal protein ERp28 endoplasmic reticulum protein 29 Endoplasmic reticulum resident protein 28 endoplasmic reticulum resident protein 29 ERp28Endoplasmic reticulum resident protein 31 ERp29 ERp31ERP28 PDIA9 PDI-DB protein disulfide isomerase family A member 9; endoplasmic reticulum lumenal protein ERp28; Endoplasmic reticulum resident protein 28; Endoplasmic reticulum resident protein 29; Endoplasmic reticulum resident protein 31; epididymis secretory protein Li 107; ERp28; ERp29; ERp31; protein disulfide isomerase family A, member 9
Gene Aliases: C12orf8; ERP28; ERP29; ERp31; HEL-S-107; PDI-DB; PDIA9
UniProt ID: (Human) P30040
Entrez Gene ID: (Human) 10961
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.