Novus Biologicals
Manufacturer Code:NBP250389AF488
Catalog # NBP250389AG
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | M37-P5D11 |
Immunogen | Peptide - LAPSRGEDDG |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DKFZp779C1042 DKFZp779I0141 EC 1.8.4 EC 1.8.4.- Endoplasmic oxidoreductin-1-like protein B ERO1-L(beta) ERO1-L-beta ERO1-like beta (S. cerevisiae) ERO1-like protein beta FLJ11003 oxidoreductin-1-L-beta; endoplasmic oxidoreductin-1-like protein B; Endoplasmic reticulum oxidoreductase beta; Endoplasmic reticulum oxidoreductin-1-like protein B; ERO1-L-beta; ERO1-like beta; ERO1-like protein beta; Oxidoreductin-1-L-beta
Gene Aliases: ERO1B; Ero1beta; ERO1LB
UniProt ID: (Human) Q86YB8
Entrez Gene ID: (Human) 56605
Molecular Function: oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.