Invitrogen
Manufacturer Code:PA5113082
Catalog # PIPA5113082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human ERO1-like protein beta protein (101-400AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1300013B24Rik 1700065B09Rik AI447560 endoplasmic oxidoreductase 1 beta endoplasmic oxidoreductin-1-like protein B endoplasmic reticulum oxidoreductase 1 beta Endoplasmic reticulum oxidoreductase beta Endoplasmic reticulum oxidoreductin-1-like protein B ERO1B Ero1beta ero1-beta Ero1lb ERO1-L-beta ERO1-like beta ERO1-like beta (S. cerevisiae) ERO1-like protein beta Oxidoreductin-1-L-beta RGD1563548; endoplasmic oxidoreductase 1 beta; endoplasmic oxidoreductin-1-like protein B; Endoplasmic reticulum oxidoreductase beta; Endoplasmic reticulum oxidoreductin-1-like protein B; ERO1-L-beta; ERO1-like beta; ERO1-like protein beta; Oxidoreductin-1-L-beta
Gene Aliases: 1300013B24Rik; 1700065B09Rik; AI447560; ero1-beta; ERO1B; Ero1beta; ERO1LB
UniProt ID: (Human) Q86YB8, (Mouse) Q8R2E9
Entrez Gene ID: (Human) 56605, (Mouse) 67475
Molecular Function: oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.