Invitrogen
Manufacturer Code:PA5116228
Catalog # PIPA5116228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 883-911 amino acids from the C-terminal region of mouse Ern2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: endoplasmic reticulum (ER) to nucleus signalling 2 endoplasmic reticulum to nucleus signaling 2 endoplasmic reticulum to nucleus signalling 2 endoplasmic reticulum-to-nucleus signaling 2 Endoribonuclease ER to nucleus signalling 2 Ern1 ern2 hIRE2p inositol-requiring 1 (Yeast homologue) inositol-requiring 1 beta inositol-requiring protein 2 Ire1 IRE1 beta IRE1 S. cerevisiae homolog of IRE1b ire1-beta Ire2 IRE2p mIre1 Serine/threonine-protein kinase Serine/threonine-protein kinase/endoribonuclease IRE2; Endoplasmic reticulum-to-nucleus signaling 2; Endoribonuclease; ern2; inositol-requiring 1 (Yeast homologue); inositol-requiring 1 beta; Inositol-requiring protein 2; Ire1-beta; IRE1b; mIre1; Serine/threonine-protein kinase; Serine/threonine-protein kinase/endoribonuclease IRE2
Gene Aliases: Ern1; Ern2; Ire1; Ire1b; Ire2
UniProt ID: (Mouse) Q9Z2E3
Entrez Gene ID: (Mouse) 26918
Molecular Function:
kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.