Novus Biologicals
Manufacturer Code:NBP183092
Catalog # NBP183092
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SPGKYMSHLASCGMGLLLTVDPGSGTVLWTQDLGVPVMGVYTWHQDGLRQLPHLTLARDTLHFLALRWGHIRLPASGPRDTATLFSTLDTQLLMTLYVGKDETGFYVSKALVHTGVALVPRGLTLAPADGPTT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 endoplasmic reticulum to nucleus signaling 2 endoplasmic reticulum to nucleus signalling 2 Endoplasmic reticulum-to-nucleus signaling 2 ER to nucleus signalling 2 hIRE2p inositol-requiring 1 beta Inositol-requiring protein 2 IRE1 S. cerevisiae homolog of Ire1-beta IRE1bIRE1 beta IRE2 serine/threonine-protein kinase/endoribonuclease IRE2; endoplasmic reticulum to nucleus signalling 2; Endoplasmic reticulum-to-nucleus signaling 2; Endoribonuclease; ER to nucleus signalling 2; hIRE2p; inositol-requiring 1 beta; Inositol-requiring protein 2; IRE1 beta; IRE1, S. cerevisiae, homolog of; Ire1-beta; IRE1b; Serine/threonine-protein kinase; Serine/threonine-protein kinase/endoribonuclease IRE2
Gene Aliases: ERN2; hIRE2p; IRE1-BETA; IRE1b; IRE2; IRE2p
UniProt ID: (Human) Q76MJ5
Entrez Gene ID: (Human) 10595
Molecular Function: kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.