Abnova
Manufacturer Code:PAB2676
Catalog # 89-128-401
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human ERN2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ERN2.
For Research Use Only
Protein Aliases: endoplasmic reticulum to nucleus signalling 2; Endoplasmic reticulum-to-nucleus signaling 2; Endoribonuclease; ER to nucleus signalling 2; hIRE2p; inositol-requiring 1 beta; Inositol-requiring protein 2; IRE1 beta; IRE1, S. cerevisiae, homolog of; Ire1-beta; IRE1b; Serine/threonine-protein kinase; Serine/threonine-protein kinase/endoribonuclease IRE2
Gene Aliases: ERN2; hIRE2p; IRE1-BETA; IRE1b; IRE2; IRE2p
UniProt ID: (Human) Q76MJ5
Entrez Gene ID: (Human) 10595
Molecular Function:
kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.