Invitrogen
Manufacturer Code:PA5115434
Catalog # PIPA5115434
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human IRE1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: endoplasmic reticulum (ER) to nucleus signalling 1; endoplasmic reticulum (ER) to nucleus signalling 2; endoplasmic reticulum to nucleus signalling 2; Endoplasmic reticulum-to-nucleus signaling 1; Endoplasmic reticulum-to-nucleus signaling 2; Endoribonuclease; ER to nucleus signalling 1; ER to nucleus signalling 2; ERN1; ERN1 ERN2 IRE1 IRE2; ern2; hIRE1p; hIRE2p; inositol-requiring 1; inositol-requiring 1 (Yeast homologue); inositol-requiring 1 alpha; inositol-requiring 1 beta; inositol-requiring enzyme 1; Inositol-requiring protein 1; Inositol-requiring protein 2; IRE1 beta; IRE1, S. cerevisiae, homolog of; Ire1-alpha; Ire1-beta; IRE1a; IRE1b; mIre1; protein kinase/endoribonuclease; Serine/threonine-protein kinase; Serine/threonine-protein kinase/endoribonuclease IRE1; Serine/threonine-protein kinase/endoribonuclease IRE2
Gene Aliases: 9030414B18Rik; AI225830; C85377; ERN1; ERN2; hIRE1p; hIRE2p; IRE1; IRE1-BETA; IRE1a; Ire1alpha; IRE1b; IRE1P; IRE2; IRE2p; RGD1559716
UniProt ID: (Human) O75460, (Human) Q76MJ5, (Mouse) Q9EQY0, (Mouse) Q9Z2E3
Entrez Gene ID: (Human) 2081, (Human) 10595, (Mouse) 78943, (Rat) 498013, (Mouse) 26918, (Rat) 365363
Molecular Function:
kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.