Novus Biologicals
Manufacturer Code:NBP213167AF700
Catalog # NBP213167Z
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3C2 |
Immunogen | Full length human recombinant ERManI [Swiss-Prot# Q9UKM7] |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha 12-mannosidase EC 3.2.1.113 endoplasmic reticulum alpha-mannosidase 1 endoplasmic reticulum mannosyl-oligosaccharide 12-alpha-mannosidase endoplasmic reticulum mannosyl-oligosaccharide 12-alpha-mannosidase 1 ER alpha 12-mannosidase ER alpha-12-mannosidase ER ManI ER mannosidase 1 ERMan1 Man9GlcNAc2-specific processing alpha-mannosidase Man9GlcNAc2-specific-processing alpha-mannosidase MANA-ER Mannosidase alpha class 1B member 1 mannosidase alpha class 1B member 1 MRT15 RP11-350O14.2; endoplasmic Reticulum Class I alpha-mannosidase; Endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase; endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase 1; ER alpha 1,2-mannosidase; ER alpha-1,2-mannosidase; ER mannosidase 1; ERMan1; Man9GlcNAc2-specific processing alpha-mannosidase; Man9GlcNAc2-specific-processing alpha-mannosidase; Mannosidase alpha class 1B member 1; mannosidase, alpha, class 1B, member 1
Gene Aliases: ERMAN1; ERManI; MAN1B1; MANA-ER; MRT15; UNQ747/PRO1477
UniProt ID: (Human) Q9UKM7
Entrez Gene ID: (Human) 11253
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.