Proteintech
Manufacturer Code:666051IG150UL
Catalog # 50-173-7023
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B1C1 |
Immunogen | ERMN Fusion Protein Ag27040 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Ermin; ermin ermin ERM like protein ERMN JN Juxtanodin KIAA1189; ermin, ERM-like protein; JN; Juxtanodin
Gene Aliases: A330104H05Rik; AI854460; ermin; ERMN; JN; juxtanodin; KIAA1189; mKIAA1189; RGD1308367
UniProt ID: (Human) Q8TAM6, (Mouse) Q5EBJ4, (Rat) Q5RJL0
Entrez Gene ID: (Pig) 100627261, (Human) 57471, (Mouse) 77767, (Rat) 295619
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.