Invitrogen
Manufacturer Code:PA544502
Catalog # PIPA544502
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human KIAA1189 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ermin; ermin, ERM-like protein; JN; Juxtanodin
Gene Aliases: ERMN; JN; juxtanodin; KIAA1189; RGD1308367
UniProt ID: (Human) Q8TAM6, (Rat) Q5RJL0
Entrez Gene ID: (Dog) 607626, (Rabbit) 100353598, (Human) 57471, (Rat) 295619
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.