Novus Biologicals
Manufacturer Code:NBP156953
Catalog # NBP156953
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to KIAA1189 The peptide sequence was selected from the middle region of KIAA1189. Peptide sequence RVIEFKKKHEEVSQFKEEGDASEDSPLSSASSQAVTPDEQPTLGKKSDIS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ermin; ERMIN ermin ERM-like protein JNKIAA1189ermin juxtanodin; ermin, ERM-like protein; JN; Juxtanodin
Gene Aliases: ERMN; JN; KIAA1189
UniProt ID: (Human) Q8TAM6
Entrez Gene ID: (Human) 57471
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.