R&D Systems
Manufacturer Code:MAB49281
Catalog # MAB49281
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 767505 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human ERMAP His30-Ala155 Accession # Q96PL5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: erythroblast membrane-associated protein (Scianna blood group); erythroblast membrane-associated protein erythroblast membrane-associated protein (RD and SC blood groups) erythroblast membrane-associated protein (Scianna blood group) erythroid membrane-associated protein hERMAP MGC118810 MGC118811 PRO2801 Radin blood group Radin blood group (Rd) Radin blood group antigen RD RDMGC118812 SC Scianna blood group Scianna blood group (Sc) Scianna blood group antigen SCMGC118813; Erythroid membrane-associated protein; hERMAP; Radin blood group (Rd); Radin blood group antigen; Scianna blood group (Sc); Scianna blood group antigen
Gene Aliases: BTN5; ERMAP; PRO2801; RD; SC
UniProt ID: (Human) Q96PL5
Entrez Gene ID: (Human) 114625
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.