Proteintech
Manufacturer Code:213751AP150UL
Catalog # 50-173-3392
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ERMAP Fusion Protein Ag15899 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ERMAP hERMAP PRO2801 Radin blood group antigen RD SC Scianna blood group antigen; erythroblast membrane-associated protein (Scianna blood group); Erythroid membrane-associated protein; hERMAP; Radin blood group (Rd); Radin blood group antigen; Scianna blood group (Sc); Scianna blood group antigen
Gene Aliases: BTN5; ERMAP; PRO2801; RD; SC
UniProt ID: (Human) Q96PL5
Entrez Gene ID: (Human) 114625
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.