Novus Biologicals
Manufacturer Code:NBP271243APC
Catalog # NB271243APC
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3H2 |
Immunogen | Full length recombinant protein of human ERK5/BMK1 (NP_002740) produced in HEK293T cells. |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Big MAP kinase 1; Big MAP kinase 1 BMK-1 BMK1 kinase BMK1EC 2.7.11.24 EC 2.7.11 ERK-5 ERK5MAPK 7 Extracellular signal-regulated kinase 5 extracellular-signal-regulated kinase 5 MAP kinase 7 mitogen-activated protein kinase 7 PRKM7ERK4; BMK-1; BMK1 kinase; ERK-5; Extracellular signal-regulated kinase 5; extracellular-signal-regulated kinase 5; MAP kinase 7; MAPK 7; Mitogen-activated protein kinase 7
Gene Aliases: BMK1; ERK4; ERK5; MAPK7; PRKM7
UniProt ID: (Human) Q13164
Entrez Gene ID: (Human) 5598
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.