OriGene
Manufacturer Code:CF500077
Catalog # 50-165-9346
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5E1 |
Immunogen | Recombit protein expressed in E.coli corresponding to amino acids 345-721 of human ERK3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ERK-3; ERK3 HsT17250 PRKM6 p97MAPK; Extracellular signal-regulated kinase 3; extracellular signal-regulated kinase, p97; MAP kinase 6; MAP kinase isoform p97; MAPK 6; Mitogen-activated protein kinase 6; p97-MAPK; protein kinase, mitogen-activated 5; protein kinase, mitogen-activated 6
Gene Aliases: ERK3; HsT17250; MAPK6; p97MAPK; PRKM6
UniProt ID: (Human) Q16659
Entrez Gene ID: (Human) 5597
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.