Invitrogen
Manufacturer Code:PA579632
Catalog # PIPA579632
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human MAPK6 recombit protein (Position: E520-N721). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ERK-3; Erk-3 related; Extracellular signal-regulated kinase 3; extracellular signal-regulated kinase, p97; MAP kinase 6; MAP kinase isoform p97; MAPK 6; mitogen activated protein kinase 4; mitogen activated protein kinase 6; Mitogen-activated protein kinase 6; Mitogen-activated protein kinase 6 MAP kinase 6 MAPK 6 Extracellular signal-regulated kinase 3 ERK-3 MAP kinase isoform p97 p97-MAPK MAPK6 ERK3 PRKM6; p55-MAPK; p97-MAPK; protein kinase, mitogen activated kinase 4; protein kinase, mitogen-activated 5; protein kinase, mitogen-activated 6
Gene Aliases: 2610021I23Rik; D130053K17Rik; ERK3; HsT17250; Mapk4; MAPK6; Mapk63; p97MAPK; Prkm4; PRKM6
UniProt ID: (Human) Q16659, (Mouse) Q61532, (Rat) P27704
Entrez Gene ID: (Human) 5597, (Mouse) 50772, (Rat) 58840
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.