R&D Systems
Manufacturer Code:MAB1230SP
Catalog # MAB1230SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 191801 |
Immunogen | <EM>E. coli</EM>-derived recombinant human ERK2 Met1-Ser360 Accession # Q1HBJ4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.24 ERK ERK-2 ERK2MAP kinase isoform p42 ERT1 Extracellular signal-regulated kinase 2 MAP kinase 1 MAP kinase 2 MAPK 1 MAPK1 MAPK2 mitogen-activated protein kinase 1 Mitogen-activated protein kinase 2 p38 p40 p41 p41mapk p42-MAPK P42MAPK PRKM1 PRKM1MAPK 2 PRKM2 protein tyrosine kinase ERK2; ERK-2; ERT1; Extracellular signal-regulated kinase 2; MAP kinase 1; MAP kinase 2; MAP kinase isoform p42; MAPK 2; Mitogen-activated protein kinase 1; Mitogen-activated protein kinase 2; p42-MAPK; protein tyrosine kinase ERK2
Gene Aliases: ERK; ERK-2; ERK2; ERT1; MAPK1; MAPK2; p38; p40; p41; p41mapk; p42-MAPK; P42MAPK; PRKM1; PRKM2
UniProt ID: (Human) P28482
Entrez Gene ID: (Human) 5594
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.