Invitrogen
Manufacturer Code:PA518329
Catalog # PIPA518329
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (CAAGPEMVRGQVF) corresponding to the internal amino acids of MAPK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ERK-2; ERT1; Extracellular signal-regulated kinase 2; MAP kinase 1; MAP kinase 2; MAP kinase isoform p42; MAPK 2; MAPK1 ERK2 ERK p38 p40 p41 ERT1 MAPK2 PRKM1 PRKM2 P42MAPK p41mapk mitogen-activated protein kinase 1 protein tyrosine kinase ERK2 mitogen-activated protein kinase 2 extracellular signal-regulated kinase 2 extracellular signal-regulated kinase-2; Mitogen-activated protein kinase 1; Mitogen-activated protein kinase 2; p42-MAPK; protein tyrosine kinase ERK2
Gene Aliases: ERK; ERK-2; ERK2; ERT1; MAPK1; MAPK2; p38; p40; p41; p41mapk; p42-MAPK; P42MAPK; PRKM1; PRKM2
UniProt ID: (Human) P28482
Entrez Gene ID: (Human) 5594
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.