Invitrogen
Manufacturer Code:LFMA0134
Catalog # AML0134
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 9B3 |
Immunogen | Synthetic peptide KLH coupled corresponding to the residues around Thr202/Tyr204 of human p44 MAP kinase (Erk1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ERK-1; ERK-2; ERT1; ERT2; Extracellular signal-regulated kinase 1; Extracellular signal-regulated kinase 2; extracellular signal-related kinase 1; Insulin-stimulated MAP2 kinase; MAP kinase 1; MAP kinase 2; MAP kinase 3; MAP kinase isoform p42; MAP kinase isoform p44; MAPK 1; MAPK 2; Microtubule-associated protein 2 kinase; Mitogen-activated protein kinase 1; Mitogen-activated protein kinase 1 MAP kinase 1 MAPK 1 ERT1 Extracellular signal-regulated kinase 2 ERK-2 MAP kinase isoform p42 p42-MAPK Mitogen-activated protein kinase 2 MAP kinase 2 MAPK 2 MAPK1 ERK2 PRKM1 PRKM2 Mitogen-activated protein kinase 3 MAP kinase 3 MAPK 3 ERT2 Extracellular signal-regulated kinase 1 ERK-1 Insulin-stimulated MAP2 kinase MAP kinase isoform p44 p44-MAPK Microtubule-associated protein 2 kinase p44-ERK1 MAPK3 ERK1 PRKM3 p44 p42; Mitogen-activated protein kinase 2; Mitogen-activated protein kinase 3; p42-MAPK; p44-ERK1; p44-MAPK; protein tyrosine kinase ERK2
Gene Aliases: ERK; ERK-1; ERK-2; ERK1; ERK2; ERT1; ERT2; HS44KDAP; HUMKER1A; MAPK1; MAPK2; MAPK3; p38; p40; p41; p41mapk; p42-MAPK; P42MAPK; p44-ERK1; p44-MAPK; P44ERK1; P44MAPK; PRKM1; PRKM2; PRKM3
UniProt ID: (Human) P28482, (Human) P27361
Entrez Gene ID: (Human) 5594, (Human) 5595
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.