Invitrogen
Manufacturer Code:82380
Catalog # PI82380
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a sequence in the C-terminus of rat p44 MAP Kinase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ERK-1; ERK-2; ERK2; ERT1; ERT2; Extracellular signal-regulated kinase 1; Extracellular signal-regulated kinase 2; extracellular signal-regulated kinase-1; extracellular signal-regulated kinase-2; extracellular signal-related kinase 1; extracellular-signal-regulated kinase 2; Insulin-stimulated MAP2 kinase; MAP kinase 1; MAP kinase 2; MAP kinase 3; MAP kinase isoform p42; MAP kinase isoform p44; MAPK 1; MAPK 2; MAPK 3; Microtubule-associated protein 2 kinase; mitogen activated protein kinase 1; mitogen-activated 3; Mitogen-activated protein kinase 1; Mitogen-activated protein kinase 1 MAP kinase 1 MAPK 1 ERT1 Extracellular signal-regulated kinase 2 ERK-2 MAP kinase isoform p42 p42-MAPK Mitogen-activated protein kinase 2 MAP kinase 2 MAPK 2 MAPK1 ERK2 PRKM1 PRKM2 Mitogen-activated protein kinase 3 MAP kinase 3 MAPK 3 ERT2 Extracellular signal-regulated kinase 1 ERK-1 Insulin-stimulated MAP2 kinase MAP kinase isoform p44 p44-MAPK Microtubule-associated protein 2 kinase p44-ERK1 MAPK3 ERK1 PRKM3 p44 p42; Mitogen-activated protein kinase 2; Mitogen-activated protein kinase 3; MNK1; p42-MAPK; p44 MAP kinase; p44-ERK1; p44-MAPK; pp42/MAP kinase; protein kinase; protein tyrosine kinase ERK2
Gene Aliases: 9030612K14Rik; AA407128; AU018647; C78273; ERK; ERK-1; ERK-2; ERK1; ERK2; ERT1; ERT2; Esrk1; fi06b09; HS44KDAP; HUMKER1A; I79_009500; Mapk; MAPK1; MAPK2; MAPK3; MNK1; Mtap2k; p38; p40; p41; p41mapk; p42-MAPK; P42MAPK; p44; p44-ERK1; p44-MAPK; P44ERK1; P44MAPK; PRKM1; PRKM2; PRKM3; wu:fi06b09; zERK1; zERK2
UniProt ID: (Human) P28482, (Bovine) P46196, (Human) P27361, (Rat) P63086, (Mouse) P63085, (Mouse) Q63844, (Rat) P21708
Entrez Gene ID: (Human) 5594, (Bovine) 327672, (Pig) 100153927, (Bovine) 531391, (Human) 5595, (Pig) 445013, (Chinese hamster) 100759514, (Rat) 116590, (Mouse) 26413, (Mouse) 26417, (Rat) 50689, (Zebrafish) 360144, (Zebrafish) 399480
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.