R&D Systems
Manufacturer Code:MAB1940
Catalog # MAB1940
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 250603 |
Immunogen | Human ERK1 synthetic peptide Met1-Val28 Accession # P27361 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 ERK-1 ERK1p44-MAPK ERT2 Extracellular signal-regulated kinase 1 extracellular signal-related kinase 1 HS44KDAP HUMKER1A Insulin-stimulated MAP2 kinase MAP kinase 1 MAP kinase 3 MAP kinase isoform p44 MAPK 1 MAPK 3 MAPK3 MGC20180 Microtubule-associated protein 2 kinase Mitogen-activated protein kinase 1 mitogen-activated protein kinase 3 p44erk1 p44-ERK1 P44MAPK PRKM3 PRKM3EC 2.7.11.24; ERK-1; ERT2; Extracellular signal-regulated kinase 1; extracellular signal-related kinase 1; Insulin-stimulated MAP2 kinase; MAP kinase 3; MAP kinase isoform p44; MAPK 1; Microtubule-associated protein 2 kinase; Mitogen-activated protein kinase 3; p44-ERK1; p44-MAPK
Gene Aliases: ERK-1; ERK1; ERT2; HS44KDAP; HUMKER1A; MAPK3; p44-ERK1; p44-MAPK; P44ERK1; P44MAPK; PRKM3
UniProt ID: (Human) P27361
Entrez Gene ID: (Human) 5595
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.