Novus Biologicals
Manufacturer Code:NBP1780310.05MG
Catalog # NBP17803105
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to the carboxy terminal end of human p44 MAP Kinase (ERK1) protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
Protein Aliases: EC 2.7.11 ERK-1 ERK1p44-MAPK ERT2 Extracellular signal-regulated kinase 1 extracellular signal-related kinase 1 HS44KDAP HUMKER1A Insulin-stimulated MAP2 kinase MAP kinase 1 MAP kinase 3 MAP kinase isoform p44 MAPK 1 MAPK 3 MGC20180 Microtubule-associated protein 2 kinase Mitogen-activated protein kinase 1 mitogen-activated protein kinase 3 p44erk1 p44-ERK1 p44mapk PRKM3EC 2.7.11.24; ERK-1; ERT2; Extracellular signal-regulated kinase 1; extracellular signal-related kinase 1; Insulin-stimulated MAP2 kinase; MAP kinase 3; MAP kinase isoform p44; MAPK 1; Microtubule-associated protein 2 kinase; Mitogen-activated protein kinase 3; p44-ERK1; p44-MAPK
Gene Aliases: ERK-1; ERK1; ERT2; HS44KDAP; HUMKER1A; MAPK3; p44-ERK1; p44-MAPK; P44ERK1; P44MAPK; PRKM3
UniProt ID: (Human) P27361
Entrez Gene ID: (Human) 5595
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.