Invitrogen
Manufacturer Code:PA517141
Catalog # PIPA517141
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the sequence of mouse THEX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3 ' exoribonuclease; 3&prime exoribonuclease 3&prime-5&prime exonuclease ERI1 3&prime-5&prime exoribonuclease 1 Eri-1 homolog enhanced RNAi three prime mRNA exonuclease homolog 1 histone mRNA 3&prime end-specific exonuclease histone mRNA 3&prime-end-specific exoribonuclease histone mRNA 3&prime-exonuclease 1 protein 3&primehExo three prime histone mRNA exonuclease 1 three prime mRNA exonuclease 1; 3' exoribonuclease; 3'-5' exonuclease ERI1; 3'-5' exoribonuclease 1; enhanced RNAi protein 1; Eri-1 homolog; histone mRNA 3' end-specific exonuclease; Histone mRNA 3'-exonuclease 1; three prime histone mRNA exonuclease 1
Gene Aliases: 3'exo; 3'hexo; 3110010F15Rik; eri-1; Eri1; RGD1308378; Thex1
UniProt ID: (Rat) Q5FVR4, (Mouse) Q7TMF2
Entrez Gene ID: (Rat) 361159, (Mouse) 67276
Molecular Function: RNA binding protein esterase exoribonuclease hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.