Invitrogen
Manufacturer Code:PA5109263
Catalog # PIPA5109263
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide within C-terminal human ERGI3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310015B14Rik AV318804 C20orf47 CGI-54 D2Ucla1 dJ477O4.2 Endoplasmic reticulum-Golgi intermediate compartment protein 3 ERGIC3 ERV46 NY-BR-84 PRO0989 SDBCAG84 Serologically defined breast cancer antigen NY-BR-84 Serologically defined breast cancer antigen NY-BR-84 homolog; Endoplasmic reticulum-Golgi intermediate compartment protein 3; endoplasmic reticulum-localized protein ERp43; serologically defined breast cancer antigen 84; Serologically defined breast cancer antigen NY-BR-84
Gene Aliases: C20orf47; CGI-54; dJ477O4.2; ERGIC3; ERV46; NY-BR-84; PRO0989; SDBCAG84
UniProt ID: (Human) Q9Y282
Entrez Gene ID: (Human) 51614
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.