Invitrogen
Manufacturer Code:PA596657
Catalog # PIPA596657
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 55-310 of human ERGIC2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD14 protein; Endoplasmic reticulum-Golgi intermediate compartment protein 2
Gene Aliases: 1200009B18Rik; 4930572C01Rik; AA408438; cd002; CDA14; ERGIC2; ERV41; PTX1
UniProt ID: (Human) Q96RQ1, (Mouse) Q9CR89
Entrez Gene ID: (Human) 51290, (Mouse) 67456
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.