Invitrogen
Manufacturer Code:PA5100505
Catalog # PIPA5100505
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ERGIC2 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1200009B18Rik 4930572C01Rik AA408438 cd002 CD14 protein CDA14 Endoplasmic reticulum-Golgi intermediate compartment protein 2 ERGIC and golgi 2 Ergic2 Erv41 PTX1 RGD1310606; CD14 protein; Endoplasmic reticulum-Golgi intermediate compartment protein 2
Gene Aliases: cd002; CDA14; ERGIC2; ERV41; PTX1
UniProt ID: (Human) Q96RQ1
Entrez Gene ID: (Human) 51290
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.