Novus Biologicals
Manufacturer Code:NBP30510820UL
Catalog # NB093854
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 55-310 of human ERGIC2 (NP_057654.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cd002 CD14 protein CDA14 ERGIC and golgi 2 Erv41 MGC111152 PTX1endoplasmic reticulum-Golgi intermediate compartment protein 2; CD14 protein; Endoplasmic reticulum-Golgi intermediate compartment protein 2
Gene Aliases: cd002; CDA14; ERGIC2; ERV41; PTX1
UniProt ID: (Human) Q96RQ1
Entrez Gene ID: (Human) 51290
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.