Invitrogen
Manufacturer Code:MA526245
Catalog # PIMA526245
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4H7 |
Immunogen | Full length human recombit protein of ERG produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ERG v-ets erythroblastosis virus E26 oncogene like (avian) HGNC:3446 erg-3 p55 transcriptional regulator ERG (transforming protein ERG) v-ets avian erythroblastosis virus E26 oncogene related v-ets erythroblastosis virus E26 oncogene like v-ets erythroblastosis virus E26 oncogene homolog (avian) OTTHUMP00000115970 OTTHUMP00000208036 TMPRSS2-ERG prostate cancer specific ets-related; erythroblast transformation-specific transcription factor ERG variant 10; ets-related; TMPRSS2/ERG fusion; Transcriptional regulator ERG; transcriptional regulator ERG (transforming protein ERG); Transforming protein ERG; v-ets avian erythroblastosis virus E26 oncogene homolog; v-ets avian erythroblastosis virus E26 oncogene related; v-ets erythroblastosis virus E26 oncogene homolog; v-ets erythroblastosis virus E26 oncogene like
Gene Aliases: D030036I24Rik; ERG; Erg-3; p55
UniProt ID: (Human) P11308, (Mouse) P81270
Entrez Gene ID: (Human) 2078, (Mouse) 13876
Molecular Function:
nucleic acid binding
signaling molecule
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.