Novus Biologicals
Manufacturer Code:NBP297609DL594
Catalog # NB045596
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human ERCC6L fragment. |
Conjugate | DyLight 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent helicase ERCC6-like; ATP-dependent helicase ERCC6-like DNA excision repair protein ERCC-6-like EC 3.6.1 EC 3.6.4.12 excision repair cross-complementing rodent repair deficiency complementationgroup 6-like excision repair protein ERCC6-like FLJ20105 MGC131695 PICHexcision repair cross-complementing rodent repair deficiency complementationgroup 6 - like PLK1-interacting checkpoint helicase SNF2/RAD54 family protein Tumor antigen BJ-HCC-15; DNA excision repair protein ERCC-6-like; excision repair cross-complementation group 6 like; excision repair cross-complementation group 6-like; excision repair cross-complementing rodent repair deficiency complementation group 6 - like; excision repair cross-complementing rodent repair deficiency, complementation group 6-like; excision repair protein ERCC6-like; PLK1-interacting checkpoint helicase; SNF2/RAD54 family protein; Tumor antigen BJ-HCC-15
Gene Aliases: ERCC6L; PICH; RAD26L
UniProt ID: (Human) Q2NKX8
Entrez Gene ID: (Human) 54821
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.