Proteintech
Manufacturer Code:205481AP150UL
Catalog # 50-173-2953
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARMD5 ATP dependent helicase ERCC6 CKN2 Cockayne syndrome protein CSB COFS COFS1 CSB ERCC6 RAD26; ATP-dependent helicase ERCC6; Chimeric CSB-PGBD3 protein; Chimeric ERCC6-PGBD3 protein; Cockayne syndrome group B protein; Cockayne syndrome protein CSB; CS group B correcting; DNA excision repair protein ERCC-6; ERCC6-PGBD3 fusion protein (isoform 1); excision repair cross-complementation group 6; excision repair cross-complementing rodent repair deficiency, complementation group 6
Gene Aliases: 4732403I04; ARMD5; C130058G22Rik; CKN2; COFS; COFS1; CSB; ERCC6; POF11; RAD26; UVSS1
UniProt ID: (Human) P0DP91, (Mouse) F8VPZ5
Entrez Gene ID: (Human) 2074, (Rat) 306274, (Mouse) 319955
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.