Invitrogen
Manufacturer Code:PA554007
Catalog # PIPA554007
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ERC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAST2; CAZ-associated structural protein 2; ELKS/Rab6-interacting/CAST family member 1; ERC protein 1; ERC-1; ERC1 ELKS/RAB6-interacting/CAST family member 1 Cast2 ELKS KIAA1081 MGC12974 RAB6IP2 RAB6 interacting protein 2 RAB6-interacting protein 2 Rab6-interacting protein 2 FLJ31750 OTTHUMP00000160633; RAB6 interacting protein 2; RAB6-interacting protein 2; RIM-Binding Protein
Gene Aliases: 4930404L01Rik; 5033405M01Rik; 9630025C19Rik; B430107L16Rik; CAST; CAST2; ELKS; ERC; ERC-1; ERC1; ERC1b; KIAA1081; mKIAA1081; RAB6IP2; RAB6IP2A; RAB6IP2B
UniProt ID: (Human) Q8IUD2, (Rat) Q811U3, (Mouse) Q99MI1
Entrez Gene ID: (Human) 23085, (Rat) 266806, (Mouse) 111173
Molecular Function: G-protein modulator enzyme modulator membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.