Invitrogen
Manufacturer Code:PA518105
Catalog # PIPA518105
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (YGSARSVGKVEPS) corresponding to the N-terminus amino acids of ERC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ELKS/Rab6-interacting/CAST family member 1; ERC-1; ERC1 ELKS/RAB6-interacting/CAST family member 1 Cast2 ELKS KIAA1081 MGC12974 RAB6IP2 RAB6 interacting protein 2 RAB6-interacting protein 2 Rab6-interacting protein 2 FLJ31750 OTTHUMP00000160633; RAB6 interacting protein 2; Rab6-interacting protein 2
Gene Aliases: Cast2; ELKS; ERC-1; ERC1; KIAA1081; RAB6IP2
UniProt ID: (Human) Q8IUD2
Entrez Gene ID: (Human) 23085
Molecular Function: G-protein modulator enzyme modulator membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.