OriGene
Manufacturer Code:CF503689
Catalog # 50-166-2321
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2C6 |
Immunogen | Full length human recombit protein of human ERAS produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: E-Ras; Embryonic stem cell-expressed Ras; GTPase ERas; HRAS2 HRASP; small GTPase protein E-Ras; v-Ha-ras Harvey rat sarcoma viral oncogene homolog 2; v-Ha-ras Harvey rat sarcoma viral oncogene homolog pseudogene
Gene Aliases: ERAS; HRAS2; HRASP
UniProt ID: (Human) Q7Z444
Entrez Gene ID: (Human) 3266
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.