Invitrogen
Manufacturer Code:PA581382
Catalog # PIPA581382
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the center region of the Mouse ERAS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E-Ras; Embryonic stem cell-expressed Ras; GTPase ERas; HRAS2 HRASP E-Ras GTPase ERas embryonic stem cell-expressed Ras small GTPase protein E-Ras v-Ha-ras Harvey rat sarcoma viral oncogene homolog 2 v-Ha-ras Harvey rat sarcoma viral oncogene homolog pseudogene; small GTPase E-Ras
Gene Aliases: ecat5; Eras; Ha-Ras2; HRAS2; HRasp
UniProt ID: (Mouse) Q7TN89
Entrez Gene ID: (Mouse) 353283
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.