Invitrogen
Manufacturer Code:PA541800
Catalog # PIPA541800
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human ERAL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CEGA; Conserved ERA-like GTPase; era (E. coli G-protein homolog)-like 1; Era (G-protein)-like 1; Era G-protein-like 1; Era-like 1; Era-like 12S mitochondrial rRNA chaperone 1; ERA-like GTPase; ERA-like protein 1; ERA-W; GTP-binding protein era homolog; GTPase Era, mitochondrial; GTPase, human homolog of E. coli essential cell cycle protein Era; H-ERA; M-ERA
Gene Aliases: 2610524P08Rik; 9130407C09Rik; AU019798; CEGA; ERA; ERA-W; ERAL1; ERAL1A; H-ERA; HERA; HERA-A; HERA-B; M-ERA; Mera; MERA-S; MERA-W
UniProt ID: (Human) O75616, (Bovine) A5PK43, (Mouse) Q9CZU4, (Rat) Q5EBA0
Entrez Gene ID: (Dog) 480629, (Human) 26284, (Horse) 100059531, (Rabbit) 100348774, (Bovine) 523344, (Guinea pig) 100721797, (Mouse) 57837, (Rat) 363646
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.