Novus Biologicals
Manufacturer Code:NBP270871JF646
Catalog # N270871J646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI10B4 |
Immunogen | Full length recombinant protein of human ERAB (NP_004484) produced in HEK293T cells. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 17-beta-estradiol 17-dehydrogenase; 17-beta-HSD 10 17-beta-hydroxysteroid dehydrogenase 10 17b-HSD10 3-hydroxy-2-methylbutyryl-CoA dehydrogenase ABAD amyloid-beta peptide binding alcohol dehydrogenase CAMR DUPXp11.22 EC 1.1.1.178 EC 1.1.1.35 Endoplasmic reticulum-associated amyloid beta-peptide-binding protein ERAB3-hydroxyacyl-CoA dehydrogenase type-2 HADH2AB-binding alcohol dehydrogenase hydroxyacyl-Coenzyme A dehydrogenase type II hydroxyacyl-Coenzyme Adehydrogenase type II hydroxysteroid (17-beta) dehydrogenase 10 mental retardation X-linked syndromic 10 MHBD Mitochondrial ribonuclease P protein 23-hydroxyacyl-CoA dehydrogenase type II Mitochondrial RNase P protein 2 MRPP2HCD2 MRX17 MRX31 MRXS10 SCHAD SDR5C1 short chain dehydrogenase/reductase family 5C member 1 short chain L-3-hydroxyacyl-CoA dehydrogenase type 2 short chain type dehydrogenase/reductase XH98G2 Short-chain type dehydrogenase/reductase XH98G2 Type II HADH XH98G2; 2-methyl-3-hydroxybutyryl-CoA dehydrogenase; 3-alpha-(17-beta)-hydroxysteroid dehydrogenase (NAD(+)); 3-hydroxy-2-methylbutyryl-CoA dehydrogenase; 3-hydroxyacyl-CoA dehydrogenase type II; 3-hydroxyacyl-CoA dehydrogenase type-2; 3alpha(or 20beta)-hydroxysteroid dehydrogenase; 7-alpha-hydroxysteroid dehydrogenase; AB-binding alcohol dehydrogenase; amyloid-beta peptide binding alcohol dehydrogenase; Endoplasmic reticulum-associated amyloid beta-peptide-binding protein; hydroxysteroid (17-beta) dehydrogenase 10; hydroxysteroid dehydrogenase 10; MHBD; Mitochondrial ribonuclease P protein 2; Mitochondrial RNase P protein 2; mitochondrial RNase P subunit 2; Short chain dehydrogenase/reductase family 5C member 1; short chain dehydrogenase/reductase family 5C, member 1; short chain L-3-hydroxyacyl-CoA dehydrogenase type 2; short chain type dehydrogenase/reductase XH98G2; Short-chain type dehydrogenase/reductase XH98G2; Type II HADH
Gene Aliases: 17b-HSD10; ABAD; CAMR; DUPXp11.22; ERAB; HADH2; HCD2; HSD17B10; MHBD; MRPP2; MRX17; MRX31; MRXS10; SCHAD; SDR5C1; XH98G2
UniProt ID: (Human) Q99714
Entrez Gene ID: (Human) 3028
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.