Novus Biologicals
Manufacturer Code:NBP234614B
Catalog # NBP234614B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | EPO104 |
Immunogen | Human eosinophils from a patient with hypereosinophilic syndrome |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 1.11.1.7 eosinophil peroxidase EPOEPER EPPEC 1.11.1 EPX-PEN; Eosinophil peroxidase; Eosinophil peroxidase heavy chain; Eosinophil peroxidase light chain; EPO
Gene Aliases: EPER; EPO; EPP; EPX; EPX-PEN; EPXD
UniProt ID: (Human) P11678
Entrez Gene ID: (Human) 8288
Molecular Function: oxidoreductase peroxidase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.