Invitrogen
Manufacturer Code:PA542062
Catalog # PIPA542062
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human EPX |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Eosinophil peroxidase; Eosinophil peroxidase heavy chain; Eosinophil peroxidase light chain; EPO
Gene Aliases: EPER; EPO; EPP; EPX; EPX-PEN; EPXD
UniProt ID: (Human) P11678, (Mouse) P49290
Entrez Gene ID: (Horse) 100070907, (Human) 8288, (Rabbit) 100356393, (Guinea pig) 100720135, (Mouse) 13861, (Rat) 303414
Molecular Function:
oxidoreductase
peroxidase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.